R&D Systems
Manufacturer Code:MAB1835500
Catalog # MAB1835500
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
In vitro Assay (IV) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1D11 |
Immunogen | Bovine bone-derived TGF-beta 1 and TGF-beta 2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ARVD1 CED DPD1 LAP LDS5 RNHF TGF beta1 TGFB TGFB1 TGFB2 TGFB3 TGFbeta TGFbeta 1 TGFbeta 2 TGFbeta 3 transforming growth factor beta 1; LAP; Latency-associated peptide; prepro-transforming growth factor beta-1; TGF-beta-1; Transforming growth factor beta-1; Transforming growth factor beta-1 proprotein
Gene Aliases: CED; DPD1; LAP; TGFB; TGFB1; TGFbeta
UniProt ID: (Human) P01137
Entrez Gene ID: (Human) 7040
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.