Invitrogen
Manufacturer Code:PA5112739
Catalog # PIPA5112739
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human dTDP-D-glucose 46-dehydratase protein (143-305AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2610017J16Rik 2610025M23Rik AI648925 CATMANS dTDP-D-glucose 46-dehydratase growth-inhibiting protein 21 SDR2E1 short chain dehydrogenase/reductase family 2E member 1 TDPGD TDP-glucose 46-dehydratase Tgds; dTDP-D-glucose 4,6-dehydratase; growth-inhibiting protein 21; short chain dehydrogenase/reductase family 2E, member 1
Gene Aliases: CATMANS; SDR2E1; TDPGD; TGDS
UniProt ID: (Human) O95455
Entrez Gene ID: (Human) 23483, (Rat) 306164
Molecular Function: dehydratase epimerase/racemase isomerase lyase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.