Novus Biologicals
Manufacturer Code:NBP187064
Catalog # NBP187064
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:SSPKQPTNPYASSKAAAECFVQSYWEQYKFPVVITRSSNVYGPHQYPEKVIPKFISLLQHNRKCCIHGSGLQTRNFLYATDVVEAFLTVL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dTDP-D-glucose 4,6-dehydratase; dTDP-D-glucose 46-dehydratase EC 4.2.1.46 growth-inhibiting protein 21 P SDR2E1 short chain dehydrogenase/reductase family 2E member 1 TDPGD TDP-glucose 46-dehydratase; growth-inhibiting protein 21; short chain dehydrogenase/reductase family 2E, member 1
Gene Aliases: CATMANS; SDR2E1; TDPGD; TGDS
UniProt ID: (Human) O95455
Entrez Gene ID: (Human) 23483
Molecular Function: dehydratase epimerase/racemase isomerase lyase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.