Proteintech
Manufacturer Code:173101AP150UL
Catalog # 50-173-1500
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TGDS Fusion Protein Ag11276 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dTDP D glucose 46 dehydratase SDR2E1 TDP glucose 46 dehydratase TDPGD TGDS; dTDP-D-glucose 4,6-dehydratase; growth-inhibiting protein 21; short chain dehydrogenase/reductase family 2E, member 1
Gene Aliases: 2610017J16Rik; 2610025M23Rik; AI648925; CATMANS; SDR2E1; TDPGD; TGDS
UniProt ID: (Human) O95455, (Mouse) Q8VDR7
Entrez Gene ID: (Human) 23483, (Rat) 306164, (Mouse) 76355
Molecular Function:
dehydratase
epimerase/racemase
isomerase
lyase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.