Invitrogen
Manufacturer Code:MA525928
Catalog # PIMA525928
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2B4 |
Immunogen | Full length human recombit protein of TFR2 produced in HEK293T cell |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: TfR2; Transferrin receptor 2 HFE3 MGC126368 TFRC2; Transferrin receptor protein 2
Gene Aliases: HFE3; TFR2; TFRC2; Trfr2
UniProt ID: (Human) Q9UP52, (Rat) B2GUY2, (Mouse) Q9JKX3
Entrez Gene ID: (Rhesus monkey) 719240, (Human) 7036, (Rat) 288562, (Mouse) 50765
Molecular Function: receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.