OriGene
Manufacturer Code:CF806306
Catalog # 50-166-7777
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3B1 |
Immunogen | Human recombit protein fragment corresponding to amino acids 29-304 of human TFPI produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: anti-convertin; EPI; EPI LACI TFI TFPI1; Extrinsic pathway inhibitor; LACI; Lipoprotein-associated coagulation inhibitor; TFPI; Tissue factor pathway inhibitor; tissue factor pathway inhibitor (lipoprotein-associated coagulation inhibitor)
Gene Aliases: EPI; LACI; TFI; TFPI; TFPI1
UniProt ID: (Human) P10646
Entrez Gene ID: (Human) 7035
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.