Novus Biologicals
Manufacturer Code:NBP28957050UL
Catalog # NB020624
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 401 |
Immunogen | Recombinant Human TFPI protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: anti-convertin; convertin EPITFPI1LACITFI Extrinsic pathway inhibitor Lipoprotein-associated coagulation inhibitor tissue factor pathway inhibitor tissue factor pathway inhibitor (lipoprotein-associated coagulation inhibitor); EPI; Extrinsic pathway inhibitor; LACI; Lipoprotein-associated coagulation inhibitor; TFPI; Tissue factor pathway inhibitor; tissue factor pathway inhibitor (lipoprotein-associated coagulation inhibitor)
Gene Aliases: EPI; LACI; TFI; TFPI; TFPI1
UniProt ID: (Human) P10646
Entrez Gene ID: (Human) 7035
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.