Invitrogen
Manufacturer Code:PA597175
Catalog # PIPA597175
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-140 of human TFIP11. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Septin and tuftelin-interacting protein 1; STIP-1; Tuftelin-interacting protein 11; tuftelin-interacting protein 33; Tuftelin-interacting protein 39; tuftelin-interacting protein, 39 kD
Gene Aliases: 2810002G02Rik; AF097181; AW046167; bK445C9.6; hNtr1; HSPC006; NTR1; Spp382; Srr1; STIP; TFIP11; TIP33; TIP39
UniProt ID: (Human) Q9UBB9, (Mouse) Q9ERA6
Entrez Gene ID: (Human) 24144, (Mouse) 54723
Molecular Function:
RNA binding protein
mRNA processing factor
mRNA splicing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.