Novus Biologicals
Manufacturer Code:NBP140364
Catalog # NBP140364
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this product maps to a region between residue 1 and 50 of human tuftelin interacting protein 11 using the numbering given in entry NP_036275.1 (GeneID 24144). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bK445C9.6 DKFZP434B194 FLJ22086 hNtr1 NTR1 Septin and tuftelin-interacting protein 1 STIP STIP-1 TIP39Spp382 tuftelin interacting protein 11 tuftelin-interacting protein 11; Septin and tuftelin-interacting protein 1; STIP-1; Tuftelin-interacting protein 11
Gene Aliases: bK445C9.6; hNtr1; HSPC006; NTR1; Spp382; STIP; TFIP11; TIP39
UniProt ID: (Human) Q9UBB9
Entrez Gene ID: (Human) 24144
Molecular Function:
RNA binding protein
mRNA processing factor
mRNA splicing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.