Abnova
Manufacturer Code:H00024144K
Catalog # 89-210-948
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human TFIP11 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human TFIP11 peptide using ARM Technology.
For Research Use Only
Protein Aliases: FLJ22086 NTR1 TIP39 bK445C9.6 hNtr1; Septin and tuftelin-interacting protein 1; STIP-1; Tuftelin-interacting protein 11
Gene Aliases: bK445C9.6; hNtr1; HSPC006; NTR1; Spp382; STIP; TFIP11; TIP39
UniProt ID: (Human) Q9UBB9
Entrez Gene ID: (Human) 24144
Molecular Function:
RNA binding protein
mRNA processing factor
mRNA splicing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.