Invitrogen
Manufacturer Code:PA568865
Catalog # PIPA568865
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of mouse TFIIIB90 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B-related factor 1; BRF-1; BRF1 homolog, subunit of RNA polymerase III transcription initiation factor IIIB; BRF1, RNA polymerase III transcription initiation factor 90 kDa subunit; hTFIIIB90; transcription factor IIIB 90 kDa subunit; Transcription factor IIIB 90 kDa subunit TFIIIB90 hTFIIIB90 B-related factor 1 BRF-1 hBRF TAF3B2 TATA box-binding protein-associated factor RNA polymerase III subunit 2 BRF1 BRF GTF3B TAF3B2 TAF3C TF3B; zinc finger protein 36, C3H type-like 1
Gene Aliases: 2510002F24Rik; GTF3B; mTFIIIB90; TAF3C; TAFIII90; TFIIIB90
Entrez Gene ID: (Dog) 490883, (Mouse) 72308, (Rat) 299347
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.