Invitrogen
Manufacturer Code:PA538755
Catalog # PIPA538755
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the internal region of human TF3B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B - related factor 1; B-related factor 1; BRF-1; BRF1 homolog, subunit of RNA polymerase III transcription initiation factor IIIB; epididymis secretory sperm binding protein Li 76p; general transcription factor IIIB, 90kD subunit; hTFIIIB90; TAF3B2; TATA box binding protein (TBP)-associated factor 3C; TATA box binding protein (TBP)-associated factor, RNA polymerase III, GTF3B subunit 2; TATA box-binding protein-associated factor, RNA polymerase III, subunit 2; TBP - associated factor, RNA polymerase III, 90kD; TFIIIB90; Transcription factor IIIB 90 kDa subunit; Transcription factor IIIB 90 kDa subunit TFIIIB90 hTFIIIB90 B-related factor 1 BRF-1 hBRF TAF3B2 TATA box-binding protein-associated factor RNA polymerase III subunit 2 BRF1 BRF GTF3B TAF3B2 TAF3C TF3B
Gene Aliases: 2510002F24Rik; BRF; BRF-1; BRF1; CFDS; GTF3B; hBRF; HEL-S-76p; mTFIIIB90; TAF3B2; TAF3C; TAFIII90; TF3B90; TFIIIB90
UniProt ID: (Human) Q92994
Entrez Gene ID: (Human) 2972, (Mouse) 72308
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.