Novus Biologicals
Manufacturer Code:NB500156
Catalog # NB500156
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide which represented a portion of human general transcription factor II-I encoded within exons 27 and 28 (LocusLink ID 2969). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BAP-135; BAP-135135kD BAP135WBS Bruton tyrosine kinase-associated protein 135 BTKAP1 DIWS FLJ38776 FLJ56355 general transcription factor II i general transcription factor IIi general transcription factor II-I GTFII-I IB291 SPINBTK-associated protein 135 TFII-ISRF-Phox1-interacting protein WBSCR6 Williams-Beuren syndrome chromosome region 6; Bruton tyrosine kinase-associated protein 135; BTK-associated protein, 135kD; General transcription factor II-I; GTFII-I; SPIN; SRF-Phox1-interacting protein; Williams-Beuren syndrome chromosomal region 6 protein; Williams-Beuren syndrome chromosome region 6
Gene Aliases: BAP135; BTKAP1; DIWS; GTF2I; GTFII-I; IB291; SPIN; TFII-I; WBS; WBSCR6
UniProt ID: (Human) P78347
Entrez Gene ID: (Human) 2969
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.