Invitrogen
Manufacturer Code:PA580112
Catalog # PIPA580112
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived rat TFF2 recombit protein (Position: E24-Y129). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: SP; Spasmolytic polypeptide; Trefoil factor 2; trefoil factor 2 (spasmolytic protein 1); Trefoil factor 2 Spasmolytic polypeptide SP Tff2 Sml1
Gene Aliases: mSP; Sml1; SP; Tff2
UniProt ID: (Rat) Q09030
Entrez Gene ID: (Rat) 116592, (Mouse) 21785
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.