Proteintech
Manufacturer Code:136811AP150UL
Catalog # 50-559-853
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TFF2 Fusion Protein Ag4537 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: SML1 SP Spasmolysin Spasmolytic polypeptide TFF2 trefoil factor 2; SP; Spasmolysin; Spasmolytic polypeptide; spasmolytic protein 1; Trefoil factor 2
Gene Aliases: mSP; SML1; SP; TFF2
UniProt ID: (Human) Q03403
Entrez Gene ID: (Human) 7032, (Mouse) 21785
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.