Novus Biologicals
Manufacturer Code:NBP185441
Catalog # NBP185441
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:QVNSAPSPSYNGSPNSFGLGEGNASPTHPVEALPVGSDHLLPSASIQDAQQWLHRNRFSQ |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CP2-related transcriptional repressor 1; CRTR-1; CRTR1CRTR-1 LBP-9 LBP9CP2-related transcriptional repressor 1 transcription factor CP2-like 1 transcription factor CP2-like protein 1 Transcription factor LBP-9; Transcription factor CP2-like protein 1; Transcription factor LBP-9
Gene Aliases: CRTR1; LBP-9; LBP9; TFCP2L1
UniProt ID: (Human) Q9NZI6
Entrez Gene ID: (Human) 29842
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.