Proteintech
Manufacturer Code:244111AP150UL
Catalog # 50-173-4340
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TFB2M Fusion Protein Ag19687 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dimethyladenosine transferase 2, mitochondrial; h mtTFB h mtTFB2 Hkp1 hTFB2M mtTFB2 NS5ATP5 TFB2M; h-mtTFB; h-mtTFB2; HCV NS5A-transactivated protein 5; Hepatitis C virus NS5A-transactivated protein 5; house-keeping protein 1; hTFB2M; Mitochondrial 12S rRNA dimethylase 2; Mitochondrial transcription factor B2; mTFB2M; mtTFB2; S-adenosylmethionine-6-N', N'-adenosyl(rRNA) dimethyltransferase 2
Gene Aliases: Hkp1; mtTFB2; NS5ATP5; TFB2M
UniProt ID: (Human) Q9H5Q4, (Mouse) Q3TL26
Entrez Gene ID: (Human) 64216, (Mouse) 15278
Molecular Function:
RNA binding protein
RNA methyltransferase
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.