Novus Biologicals
Manufacturer Code:NBP189060
Catalog # NBP189060
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:YFMVPTQKVPSLQHFRKTEKEVIGGLCSLANIPLTPETQRDQERRIRREIANSNERRRMQSINAGFQSLKTLIPHTDGEKLSKAAILQQTAEYIFSLEQEKTRLLQQNTQLKRFIQELSGSSPKRRRAEDKDEGIGSPDIWEDEK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Activating enhancer-binding protein 4; Activating enhancer-binding protein 4 AP-4 BHLHC41 bHLHc41Class C basic helix-loop-helix protein 41 transcription factor AP-4 transcription factor AP-4 (activating enhancer binding protein 4) transcription factor AP-4 (activating enhancer-binding protein 4); bHLHc41; Class C basic helix-loop-helix protein 41; Transcription factor AP-4; transcription factor AP-4 (activating enhancer-binding protein 4)
Gene Aliases: AP-4; BHLHC41; TFAP4
UniProt ID: (Human) Q01664
Entrez Gene ID: (Human) 7023
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.