Invitrogen
Manufacturer Code:PA540326
Catalog # PIPA540326
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human TFAP2D |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: activating enhancer binding protein 2 beta-like 1; activating enhancer-binding protein 2 delta; Activating enhancer-binding protein 2-delta; AP-2 like; AP-2-delta; AP2-delta; TFAP2D; transcription factor AP-2 beta (activating enhancer binding protein 2 beta)-like 1; transcription factor AP-2 beta (activating enhancer-binding protein 2 beta)-like 1; transcription factor AP-2 beta-like 1; transcription factor AP-2 delta (activating enhancer binding protein 2 delta); Transcription factor AP-2 delta (activating enhancer binding protein 2 delta) TFAP2BL1; Transcription factor AP-2-beta-like 1; Transcription factor AP-2-delta
Gene Aliases: si:dkey-5e16.2; Tcfap2d; TFAP2BL1; TFAP2D; zgc:114196
UniProt ID: (Human) Q7Z6R9, (Mouse) Q91ZK0, (Zebrafish) Q5RJ20
Entrez Gene ID: (Horse) 100056165, (Human) 83741, (Dog) 481835, (Rabbit) 100338157, (Bovine) 539910, (Rat) 301284, (Mouse) 226896, (Zebrafish) 566838
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.