Novus Biologicals
Manufacturer Code:NBP128498
Catalog # NBP128498
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-HHQSFHYEFQHSHP corresponding to internal region according to NP_758438.2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: activating enhancer binding protein 2 beta-like 1; Activating enhancer-binding protein 2-delta; Activating enhancer-binding protein 2-delta AP-2 like AP2-delta TFAP2BL1activating enhancer binding protein 2 beta-like 1 transcription factor AP-2 beta (activating enhancer binding protein 2beta)-like 1 transcription factor AP-2 beta (activating enhancer-binding protein 2beta)-like 1 transcription factor AP-2 delta (activating enhancer binding protein 2 delta) Transcription factor AP-2-beta-like 1 transcription factor AP-2-delta; AP-2 like; AP2-delta; TFAP2D; transcription factor AP-2 beta (activating enhancer binding protein 2 beta)-like 1; transcription factor AP-2 beta (activating enhancer-binding protein 2 beta)-like 1; transcription factor AP-2 delta (activating enhancer binding protein 2 delta); Transcription factor AP-2-beta-like 1; Transcription factor AP-2-delta
Gene Aliases: TFAP2BL1; TFAP2D
UniProt ID: (Human) Q7Z6R9
Entrez Gene ID: (Human) 83741
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.