Proteintech
Manufacturer Code:145721AP150UL
Catalog # 50-172-9552
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TFAP2C Fusion Protein Ag5648 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: activating enhancer binding protein 2 gamma; Activating enhancer-binding protein 2 gamma; activator protein 2 gamma; ap 2gamma AP2 GAMMA ERF1 hAP 2g TFAP2C TFAP2G Transcription factor ERF 1; AP-2.2; AP2-gamma; estrogen receptor factor 1; stimulated by retinoic acid gene 2; Transcription factor AP-2 gamma; transcription factor AP-2 gamma (activating enhancer binding protein 2 gamma); transcription factor AP-2 gamma (activating enhancer-binding protein 2 gamma); Transcription factor ERF-1
Gene Aliases: AA409384; Ap-2.2; AP2-GAMMA; AP2gamma; ERF1; hAP-2g; Stra2; Tcfap2c; TFAP2C; TFAP2G
UniProt ID: (Human) Q92754, (Mouse) Q61312
Entrez Gene ID: (Human) 7022, (Rat) 362280, (Mouse) 21420
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.