Invitrogen
Manufacturer Code:PA5110475
Catalog # PIPA5110475
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-160 of human TFAP2B (NP_003212.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: activating enhancer binding protein 2 beta; activating enhancer-binding protein 2 beta; Activating enhancer-binding protein 2-beta; Activating enhancer-binding protein 2-beta AI606113 AP-2(beta) AP-2B AP2-B AP2-beta E130018K07Rik Tcfap2b TFAP2B Transcription factor AP-2-beta; AP2-beta; transcription factor AP-2 beta (activating enhancer binding protein 2 beta); Transcription factor AP-2-beta
Gene Aliases: AI606113; AP-2(beta); AP-2B; AP2-B; E130018K07Rik; Tcfap2b; TFAP2B
UniProt ID: (Human) Q92481, (Mouse) Q61313
Entrez Gene ID: (Human) 7021, (Mouse) 21419
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.