Abnova
Manufacturer Code:PAB3818
Catalog # 89-131-152
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human TFAP2A. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of TFAP2A.
For Research Use Only
Protein Aliases: Activating enhancer-binding protein 2-alpha; Activator protein 2; AP-2; AP-2 transcription factor; AP-2alpha; AP2-alpha; AP2TF; BOFS; TFAP2; transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha); Transcription factor AP-2-alpha
Gene Aliases: AP-2; AP-2alpha; AP2TF; BOFS; TFAP2; TFAP2A
UniProt ID: (Human) P05549
Entrez Gene ID: (Human) 7020
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.