Invitrogen
Manufacturer Code:PA568788
Catalog # PIPA568788
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of mouse TFAM |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MtTF1 TCF6 TCF6L1 TCF6L2 TCF6L3 mtTFA TCF-6 Transcription factor 6-like 2 (mitochondrial transcription factor) mitochondrial transcription factor 1 mitochondrial transcription factor A transcription factor 6; mtTFA; Testis-specific high mobility group protein; testis-specific HMG-box protein m-tsHMG; Transcription factor A, mitochondrial; TS-HMG
Gene Aliases: AI661103; Hmgts; Mttfa; Tfam; tsHMG
UniProt ID: (Rat) Q91ZW1, (Mouse) P40630
Entrez Gene ID: (Rabbit) 100358902, (Rat) 83474, (Mouse) 21780
Molecular Function: DNA binding protein HMG box transcription factor chromatin/chromatin-binding protein nucleic acid binding signaling molecule transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.