Abnova
Manufacturer Code:H00007019K
Catalog # 89-202-536
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human TFAM |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human TFAM peptide using ARM Technology.
For Research Use Only
Protein Aliases: Mitochondrial transcription factor 1; mitochondrial transcription factor A; MtTF1; MtTF1 TCF6 TCF6L1 TCF6L2 TCF6L3 mtTFA; mtTFA; TCF-6; Transcription factor 6; transcription factor 6-like 1; Transcription factor 6-like 2; transcription factor 6-like 2 (mitochondrial transcription factor); transcription factor 6-like 3; Transcription factor A, mitochondrial
Gene Aliases: MTTF1; MTTFA; TCF6; TCF6L1; TCF6L2; TCF6L3; TFAM
UniProt ID: (Human) Q00059
Entrez Gene ID: (Human) 7019
Molecular Function: DNA binding protein HMG box transcription factor chromatin/chromatin-binding protein nucleic acid binding signaling molecule transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.