Proteintech
Manufacturer Code:154471AP150UL
Catalog # 50-173-0235
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TEX101 Fusion Protein Ag7611 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cancer/testis antigen 131; Cell surface receptor NYD SP8 GTPR867 NYD SP8 PRO1884 SGRG TES101RP testis expressed 101 Testis expressed protein 101 TEX101; Cell surface receptor NYD-SP8; Scleroderma-associated autoantigen; spermatogenesis associated 44; Spermatogenesis-related gene protein; testis expressed sequence 101; Testis-expressed protein 101; testis-specific protein TES101RP
Gene Aliases: CT131; GTPR867; NYD-SP8; PRO1884; SGRG; SPATA44; TES101RP; TEX101; UNQ867/PRO1884
UniProt ID: (Human) Q9BY14
Entrez Gene ID: (Human) 83639
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.