Novus Biologicals
Manufacturer Code:NBP243552
Catalog # NBP243552
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 649 |
Immunogen | Recombinant protein encompassing a sequence within the N-terminus region of human TET2. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: FLJ20032 MGC125715 tet methylcytosine dioxygenase 2; Methylcytosine dioxygenase TET2; probable methylcytosine dioxygenase TET2; tet oncogene family member 2
Gene Aliases: KIAA1546; MDS; Nbla00191; TET2
UniProt ID: (Human) Q6N021
Entrez Gene ID: (Human) 54790
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.