Invitrogen
Manufacturer Code:PA576801
Catalog # PIPA576801
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human TET2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA1546 MDS Methylcytosine dioxygenase TET2 Nbla00191 Probable methylcytosine dioxygenase TET2 Protein Ayu17 449 Tet 2 Tet methylcytosine dioxygenase 2 Tet oncogene 2 Tet oncogene family member 2 TET2 TET2&lowbarHUMAN; Methylcytosine dioxygenase TET2; probable methylcytosine dioxygenase TET2; tet oncogene family member 2
Gene Aliases: KIAA1546; MDS; Nbla00191; TET2
UniProt ID: (Human) Q6N021
Entrez Gene ID: (Human) 54790
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.