Invitrogen
Manufacturer Code:PA572804
Catalog # PIPA572804
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TET2 antibody for an 18 amino acid peptide near the carboxy terminus of human TET2. The immunogen is within amino acids 1550 - 1600 of TET2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA1546 MDS tet methylcytosine dioxygenase 2; Methylcytosine dioxygenase TET2; probable methylcytosine dioxygenase TET2; Protein Ayu17-449; tet oncogene 2; tet oncogene family member 2
Gene Aliases: Ayu17-449; E130014J05Rik; KIAA1546; MDS; mKIAA1546; Nbla00191; TET2
UniProt ID: (Human) Q6N021, (Mouse) Q4JK59
Entrez Gene ID: (Human) 54790, (Mouse) 214133
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.