Abnova
Manufacturer Code:PAB27176
Catalog # 89-128-827
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 18 amino acids at N-terminus of human TET2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of TET2.
For Research Use Only
Protein Aliases: FLJ20032; KIAA1546; Methylcytosine dioxygenase TET2; MGC125715; probable methylcytosine dioxygenase TET2; tet oncogene family member 2
Gene Aliases: KIAA1546; MDS; Nbla00191; TET2
UniProt ID: (Human) Q6N021
Entrez Gene ID: (Human) 54790
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.