Invitrogen
Manufacturer Code:PA566211
Catalog # PIPA566211
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human TESPA1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HSPC257 KIAA0748 HSPC257 Protein TESPA1 Thymocyte-expressed positive selection-associated protein 1; Protein TESPA1; Thymocyte-expressed positive selection-associated protein 1
Gene Aliases: HSPC257; KIAA0748; TESPA1
UniProt ID: (Human) A2RU30
Entrez Gene ID: (Human) 9840
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.