Novus Biologicals
Manufacturer Code:NB100317SS
Catalog # NB100317SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2C4 |
Immunogen | Full-length recombinant human Telomerase reverse transcriptase (TERT) from insect cells. [Uniprot: O14746]. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EST2Telomerase-associated protein 2 hEST2 TCS1EC 2.7.7.49 Telomerase catalytic subunit telomerase reverse transcriptase TP2HEST2 TRTEC 2.7.7; HEST2; Telomerase catalytic subunit; Telomerase reverse transcriptase; Telomerase-associated protein 2; TP2
Gene Aliases: CMM9; DKCA2; DKCB4; EST2; hEST2; hTRT; PFBMFT1; TCS1; TERT; TP2; TRT
UniProt ID: (Human) O14746
Entrez Gene ID: (Human) 7015
Molecular Function:
RNA binding protein
nucleic acid binding
reverse transcriptase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.