Invitrogen
Manufacturer Code:PA586750
Catalog # PIPA586750
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to residues in Human TERT. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HEST2; Telomerase catalytic subunit; Telomerase reverse transcriptase; telomerase reverse transcriptase beta; Telomerase reverse transcriptase HEST2 Telomerase catalytic subunit Telomerase-associated protein 2 TP2 TERT EST2 TCS1 TRT; Telomerase-associated protein 2; tert {ECO:0000312|RGD:70494}; TP2
Gene Aliases: CMM9; DKCA2; DKCB4; EST2; hEST2; hTRT; PFBMFT1; TCS1; TERT; TP2; TR; TRT
UniProt ID: (Human) O14746, (Rat) Q673L6, (Mouse) O70372
Entrez Gene ID: (Human) 7015, (Rat) 301965, (Mouse) 21752
Molecular Function: RNA binding protein nucleic acid binding reverse transcriptase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.