Invitrogen
Manufacturer Code:PA580105
Catalog # PIPA580105
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the N-terminus of mouse TelomerasE. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Telomerase catalytic subunit; Telomerase reverse transcriptase; telomerase reverse transcriptase beta; Telomerase reverse transcriptase Telomerase catalytic subunit Tert; tert {ECO:0000312|RGD:70494}
Gene Aliases: EST2; TCS1; Tert; TP2; TR; TRT
UniProt ID: (Rat) Q673L6, (Mouse) O70372
Entrez Gene ID: (Rat) 301965, (Mouse) 21752
Molecular Function:
RNA binding protein
nucleic acid binding
reverse transcriptase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.