Invitrogen
Manufacturer Code:PA538035
Catalog # PIPA538035
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | 15 amino acid peptide near the carboxy terminus of human UBIAD1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: SCCD TERE1 UbiA prenyltransferase domain-containing protein 1 Transitional epithelial response protein 1; transitional epithelia response protein; Transitional epithelial response protein 1; UbiA prenyltransferase domain-containing protein 1
Gene Aliases: 1200002M06Rik; AI426463; AW320947; RGD1309588; SCCD; TERE1; UBIAD1
UniProt ID: (Human) Q9Y5Z9, (Rat) D3ZG27, (Mouse) Q9DC60
Entrez Gene ID: (Human) 29914, (Rat) 313706, (Mouse) 71707
Molecular Function:
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.