Invitrogen
Manufacturer Code:PA531845
Catalog # PIPA531845
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 307 of Human TENR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adenosine deaminase domain containing 1 (testis-specific); Adenosine deaminase domain-containing protein 1; Tenr adenosine deaminase domain-containing protein 1 testis nuclear RNA-binding protein; Testis nuclear RNA-binding protein; testis tissue sperm-binding protein Li 92mP
Gene Aliases: ADAD1; TENR
UniProt ID: (Human) Q96M93
Entrez Gene ID: (Human) 132612
Molecular Function:
DNA binding protein
RNA binding protein
deaminase
defense/immunity protein
enzyme modulator
hydrolase
kinase activator
kinase modulator
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.