Invitrogen
Manufacturer Code:PA521966
Catalog # PIPA521966
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1088 and 1409 of TENC1 (Uniprot IDNo.Q63HR2) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C1 domain-containing phosphatase and tensin homolog; C1-TEN; C1-TEN C1TEN TNS2 C1 domain-containing phosphatase and tensin homolog tensin 2 tensin like C1 domain-containing phosphatase tensin-2 tensin-like C1 domain-containing phosphatase; tensin like C1 domain containing phosphatase (tensin 2); tensin like C1 domain-containing phosphatase; Tensin-2; Tensin-like C1 domain-containing phosphatase
Gene Aliases: C1-TEN; C1TEN; KIAA1075; TENC1; TNS2
UniProt ID: (Human) Q63HR2
Entrez Gene ID: (Human) 23371
Molecular Function:
hydrolase
phosphatase
protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.