Abnova
Manufacturer Code:PAB8498
Catalog # 89-133-710
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to N-terminus of human TENC1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of TENC1.
For Research Use Only
Protein Aliases: C1 domain-containing phosphatase and tensin homolog; C1-TEN; C1TEN; DKFZp686D13244; FLJ16320; KIAA1075; tensin like C1 domain containing phosphatase (tensin 2); tensin like C1 domain-containing phosphatase; Tensin-2; Tensin-like C1 domain-containing phosphatase; TNS2
Gene Aliases: C1-TEN; C1TEN; KIAA1075; TENC1; TNS2
UniProt ID: (Human) Q63HR2
Entrez Gene ID: (Human) 23371
Molecular Function: hydrolase phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.