R&D Systems
Manufacturer Code:FAB8896G100UG
Catalog # FAB8896G100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 499716 |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human TEM5/GPR124 Met1-Leu699 Accession # Q96PE1 |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: Adhesion G protein-coupled receptor A2; DKFZp434C211 DKFZp434J0911 G protein-coupled receptor 124 GPR124 G-protein coupled receptor 124 KIAA1531 KIAA1531FLJ14390 TEM5Tumor endothelial marker 5; G protein-coupled receptor 124; G-protein coupled receptor 124; Tumor endothelial marker 5
Gene Aliases: ADGRA2; GPR124; KIAA1531; TEM5
UniProt ID: (Human) Q96PE1
Entrez Gene ID: (Human) 25960
Molecular Function: G-protein coupled receptor antibacterial response protein defense/immunity protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.