Invitrogen
Manufacturer Code:PA533619
Catalog # PIPA533619
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 19 amino acid peptide from N-terminal extracellular domain of human GPR124. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adhesion G protein-coupled receptor A2; G protein-coupled receptor 124; G-protein coupled receptor 124; GPR124 G protein-coupled receptor 124 G-protein coupled receptor 124 TEM5 KIAA1531 Tumor endothelial marker 5; Tumor endothelial marker 5
Gene Aliases: ADGRA2; GPR124; KIAA1531; TEM5
UniProt ID: (Human) Q96PE1
Entrez Gene ID: (Human) 25960
Molecular Function: G-protein coupled receptor antibacterial response protein defense/immunity protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.