Novus Biologicals
Manufacturer Code:NBP288428
Catalog # NB036050
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of human TEM |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BRICD4 BRICHOS domain containing 4 CHM1L chondromodulin-I-like protein hChM1L hTeM myodulin TEM tendin tenomodulin UNQ771/PRO1565; BRICHOS domain containing 4; ChM1L; Chondromodulin-1-like protein; Chondromodulin-I-like protein; chondromodulin-IB; hChM1L; hTeM; Myodulin; TeM; Tendin; Tenomodulin
Gene Aliases: BRICD4; CHM1L; TEM; TNMD; UNQ771/PRO1565
UniProt ID: (Human) Q9H2S6
Entrez Gene ID: (Human) 64102
Molecular Function: membrane-bound signaling molecule signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.