Proteintech
Manufacturer Code:660771IG150UL
Catalog # 50-173-6514
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3H10F4 |
Immunogen | TELO2 Fusion Protein Ag8834 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: hCLK2; hCLK2 KIAA0683 Protein clk 2 homolog TEL2 TELO2; Protein clk-2 homolog; TEL2, telomere maintenance 2, homolog; Telomere length regulation protein TEL2 homolog; telomere maintenance 2
Gene Aliases: CLK2; KIAA0683; RGD1305553; TEL2; TELO2; YHFS
UniProt ID: (Human) Q9Y4R8
Entrez Gene ID: (Human) 9894, (Rat) 302986
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.