Invitrogen
Manufacturer Code:PA559583
Catalog # PIPA559583
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human TELO2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CLK2 TEL2 protein clk-2 homolog telomere length regulation protein TEL2 homolog; hCLK2; Protein clk-2 homolog; TEL2, telomere maintenance 2, homolog; Telomere length regulation protein TEL2 homolog
Gene Aliases: CLK2; KIAA0683; TEL2; TELO2; YHFS
UniProt ID: (Human) Q9Y4R8
Entrez Gene ID: (Human) 9894
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.