Invitrogen
Manufacturer Code:PA5116718
Catalog # PIPA5116718
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 450-550 of human TELO2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1200003M09Rik AI415602 CLK2 hCLK2 Kiaa0683 mKIAA0683 Protein clk-2 homolog RGD1305553 TEL2 TEL2 telomere maintenance 2 homolog TEL2 telomere maintenance 2 homolog (S. cerevisiae) Telo2 Telomere length regulation protein TEL2 homolog telomere maintenance 2 YHFS; hCLK2; Protein clk-2 homolog; TEL2, telomere maintenance 2, homolog; Telomere length regulation protein TEL2 homolog
Gene Aliases: 1200003M09Rik; AI415602; CLK2; KIAA0683; mKIAA0683; TEL2; TELO2; YHFS
UniProt ID: (Human) Q9Y4R8, (Mouse) Q9DC40
Entrez Gene ID: (Human) 9894, (Mouse) 71718
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.