Invitrogen
Manufacturer Code:PA570345
Catalog # PIPA570345
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N-terminal region of mouse TEKT3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Tektin 3; Tektin-3; testicular microtubules-related protein
Gene Aliases: 4933407G07Rik; OMC45; TEKT3
UniProt ID: (Bovine) A6H782, (Human) Q9BXF9, (Mouse) Q6X6Z7, (Rat) Q4V8G8
Entrez Gene ID: (Dog) 479510, (Rabbit) 100356322, (Horse) 100073130, (Bovine) 522915, (Human) 64518, (Guinea pig) 100723525, (Mouse) 71062, (Rat) 287392
Molecular Function: cytoskeletal protein microtubule family cytoskeletal protein non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.