Novus Biologicals
Manufacturer Code:NBP174181
Catalog # NBP174181
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to the N terminal of Tekt3. Immunizing peptide sequence MLPFVSNRTTLFTRYTPDDWYRSTLVGFQESNCSRHNSERLRVDTSRLIQ. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ32828 tektin 3 tektin-3 testicular microtubules-related protein; Tektin-3; testicular microtubules-related protein
Gene Aliases: TEKT3
UniProt ID: (Human) Q9BXF9
Entrez Gene ID: (Human) 64518
Molecular Function: cytoskeletal protein microtubule family cytoskeletal protein non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.