Novus Biologicals
Manufacturer Code:NBP186955
Catalog # NBP186955
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:HQIRQEARVLRNETNNQTIWDEHDNRTRLVERIDTVNRWKEMLDKCLTDLDAEIDALTQMKESAEQNLQAKNLPLDVAIECLTLRESRRDIDVVKD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: h-tektin-t TEKTB1tektin-B1 tektin 2 (testicular) tektin-2 Tektin-B1 TEKTIN-T Testicular tektin Testicular tektin B1-like protein; TEKTB1; tektin 2 (testicular); Tektin-2; tektin-B1; Tektin-t; Testicular tektin; Testicular tektin B1-like protein
Gene Aliases: h-tektin-t; TEKT2; TEKTB1; TEKTIN-T
UniProt ID: (Human) Q9UIF3
Entrez Gene ID: (Human) 27285
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.