Invitrogen
Manufacturer Code:PA597096
Catalog # PIPA597096
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 36-210 of human TEFM. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: transcription elongation factor of mitochondria; Transcription elongation factor, mitochondrial
Gene Aliases: 1110002N22Rik; C17orf42; RGD1306595; TEFM
UniProt ID: (Human) Q96QE5, (Mouse) Q5SSK3, (Rat) Q4KM51
Entrez Gene ID: (Human) 79736, (Mouse) 68550, (Rat) 287554
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.