Novus Biologicals
Manufacturer Code:NBP255665
Catalog # NBP255665
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:GSVLFTAGERWRCFLTPSRSSLYWALHNFCCRKKSTTPKKITPNVTFCDENAKEPENALDKLFSSEQQASILHVLNTASTKELEAFRLLRG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C17orf42 chromosome 17 open reading frame 42 FLJ22729 MGC24674 TEFM transcription elongation factor of mitochondria transcription elongation factor mitochondrial; transcription elongation factor of mitochondria; Transcription elongation factor, mitochondrial
Gene Aliases: C17orf42; TEFM
UniProt ID: (Human) Q96QE5
Entrez Gene ID: (Human) 79736
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.