Novus Biologicals
Manufacturer Code:NBP29408020UL
Catalog # NB022720
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 36-210 of human TEFM (NP_078959.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C17orf42 chromosome 17 open reading frame 42 FLJ22729 MGC24674 TEFM transcription elongation factor of mitochondria transcription elongation factor mitochondrial; transcription elongation factor of mitochondria; Transcription elongation factor, mitochondrial
Gene Aliases: C17orf42; TEFM
UniProt ID: (Human) Q96QE5
Entrez Gene ID: (Human) 79736
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.